Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetylphenoxyacetic acid, 98+%

$54.85 - $453.54

Chemical Identifiers

Chemical Name or Material 4-Acetylphenoxyacetic acid
CAS 1878-81-5
Molecular Formula C10H10O4
Formula Weight 194.19
MDL Number MFCD00014363
Beilstein 1956136
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0042606

  SDS
alfa aesar™
L0042606
5g Each for $54.85

AAL0042614

  SDS
alfa aesar™
L0042614
25g Each for $168.76

AAL0042622

  SDS
alfa aesar™
L0042622
100g Each for $453.54
Description & Specifications

Specifications

Formula Weight 194.19
CAS 1878-81-5
MDL Number MFCD00014363
Chemical Name or Material 4-Acetylphenoxyacetic acid
Beilstein 1956136
Molecular Formula C10H10O4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 172° to 174°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Assay ≥98%
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.