missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetylindan 98.0+%, TCI America™

$57.44 - $284.28

Chemical Identifiers

CAS 38997-97-6
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.216
MDL Number MFCD09033715
InChI Key FPHUGFGBPNVMQR-UHFFFAOYSA-N
PubChem CID 13657961
IUPAC Name 1-(2,3-dihydro-1H-inden-4-yl)ethanone
SMILES CC(=O)C1=C2CCCC2=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A22961G
View Documents
Tci America
A22961G
1g
Each for $57.44
 
A22965G
View Documents
Tci America
A22965G
5g
Each for $284.28
 
Description

Description

Specifications

Chemical Identifiers

38997-97-6
160.216
FPHUGFGBPNVMQR-UHFFFAOYSA-N
1-(2,3-dihydro-1H-inden-4-yl)ethanone
C11H12O
MFCD09033715
13657961
CC(=O)C1=C2CCCC2=CC=C1

Specifications

103°C
41°C
38997-97-6
MFCD09033715
CC(=O)C1=C2CCCC2=CC=C1
160.216
160.22
≥98.0% (GC)
White-Yellow
1g
C11H12O
FPHUGFGBPNVMQR-UHFFFAOYSA-N
1-(2,3-dihydro-1H-inden-4-yl)ethanone
13657961
Crystal-Lump at 20°C
4-Acetylindan
Safety and Handling

Safety and Handling

TSCA : No

SDS