Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetyldiphenyl sulfone, 98%

$144.20 - $144.20

Chemical Identifiers

Chemical Name or Material 4-Acetyldiphenyl sulfone
CAS 65085-83-8
Molecular Formula C14H12O3S
Formula Weight 260.31
MDL Number MFCD00209628
Synonym 4'-(Phenylsulfonyl)acetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1837603

  SDS
alfa aesar™
A1837603
1g Each for $144.20
Description & Specifications

Specifications

Formula Weight 260.31
Chemical Name or Material 4-Acetyldiphenyl sulfone
Quantity 1g
CAS 65085-83-8
Assay 98%
Synonym 4'-(Phenylsulfonyl)acetophenone
Molecular Formula C14H12O3S
MDL Number MFCD00209628
Melting Point 138° to 140°C

Applications
4-Acetyldiphenyl sulfone is used as an intermediate in organic synthesis.

Notes
Incompatible with strong oxidizing agents.