Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetyldiphenyl sulfide, 97%

$76.39 - $76.39

Chemical Identifiers

Chemical Name or Material 4-Acetyldiphenyl sulfide
CAS 10169-55-8
Molecular Formula C14H12OS
Formula Weight 228.32
MDL Number MFCD00026227
Beilstein 2049296
Synonym 4'-(Phenylthio)acetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2007303

  SDS
alfa aesar™
B2007303
1g Each for $76.39
Description & Specifications

Specifications

Formula Weight 228.32
Chemical Name or Material 4-Acetyldiphenyl sulfide
Assay 97%
Molecular Formula C14H12OS
MDL Number MFCD00026227
CAS 10169-55-8
Beilstein 2049296
Synonym 4'-(Phenylthio)acetophenone
Boiling Point 183° to 184°C (3mmHg)
Melting Point 64° to 68°C
Quantity 1g
Odor Odorless