Learn More
4-Acetylbiphenyl, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1462214
Description
4-Acetylbiphenyl is involved in the study of inactivation of 7-ethoxy-4-trifluoromethylcoumarin ortho-deethylase activity by various arylalkynes. Further, it is used in the preparation of Schiffs base.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Acetylbiphenyl is involved in the study of inactivation of 7-ethoxy-4-trifluoromethylcoumarin ortho-deethylase activity by various arylalkynes. Further, it is used in the preparation of Schiffs base.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
4-Acetylbiphenyl | |
117°C to 123°C | |
C14H12O | |
25 g | |
4-acetylbiphenyl, 4'-phenylacetophenone, 4-phenylacetophenone, 4-biphenylyl methyl ketone, p-phenylacetophenone, p-acetylbiphenyl, 1-1,1'-biphenyl-4-yl ethanone, acetophenone, 4'-phenyl, 1-4-phenylphenyl ethanone, ketone, 4-biphenylyl methyl | |
QCZZSANNLWPGEA-UHFFFAOYSA-N | |
1-(4-phenylphenyl)ethanone | |
7113 | |
98% |
92-91-1 | |
168°C (8 mmHg) | |
MFCD00008749 | |
1101615 | |
Insoluble in water. | |
CC(=O)C1=CC=C(C=C1)C2=CC=CC=C2 | |
196.249 | |
196.25 |
Chemical Identifiers
92-91-1 | |
196.249 | |
QCZZSANNLWPGEA-UHFFFAOYSA-N | |
7113 | |
CC(=O)C1=CC=C(C=C1)C2=CC=CC=C2 |
C14H12O | |
MFCD00008749 | |
4-acetylbiphenyl, 4'-phenylacetophenone, 4-phenylacetophenone, 4-biphenylyl methyl ketone, p-phenylacetophenone, p-acetylbiphenyl, 1-1,1'-biphenyl-4-yl ethanone, acetophenone, 4'-phenyl, 1-4-phenylphenyl ethanone, ketone, 4-biphenylyl methyl | |
1-(4-phenylphenyl)ethanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 202-202-6
RTECSNumber : AM9662502
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only