Please login to your online account to display your discounted pricing

4-Acetylbiphenyl 97%, ACROS Organics™


Quantity 250g

Manufacturer: thermo fisher scientific  153662500

 View more versions of this product

Catalog No. AC153662500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 196.25
Quantity 250g
Color Off-white to beige
EINECS Number 202-202-6
RTECS Number AM9662502
TSCA TSCA
Melting Point 118° to 123°C
CAS Min % 96.0
CAS Max % 100.0
Name Note 97%
Synonym 4-Phenylacetophenone
Chemical Name or Material 4-Acetylbiphenyl
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C14H12O
Linear Formula C6H5C6H4COCH3
MDL Number MFCD00008749
CAS 92-91-1

Specifications

Chemical Name or Material 4-Acetylbiphenyl
Name Note 97%
CAS 92-91-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C14H12O
Linear Formula C6H5C6H4COCH3
Formula Weight 196.25
MDL Number MFCD00008749
Synonym 4-Phenylacetophenone