Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetylbenzonitrile, 99%

Chemical Identifiers

Chemical Name or Material 4-Acetylbenzonitrile
CAS 1443-80-7
Molecular Formula C9H7NO
Formula Weight 145.16
MDL Number MFCD00001825
UN Number UN3439
Beilstein 1932887
Synonym 4'-Cyanoacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1427106

 
alfa aesar™
A1427106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1427122

 
alfa aesar™
A1427122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 145.16
Chemical Name or Material 4-Acetylbenzonitrile
CAS 1443-80-7
Boiling Point 95° to 96°C (14mmHg)
UN Number UN3439
MDL Number MFCD00001825
Melting Point 55° to 58°C
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Beilstein 1932887
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
Synonym 4'-Cyanoacetophenone
Quantity 5g
Molecular Formula C9H7NO

Applications
4-Acetylbenzonitrile is used in the preparation of antimalarial isonitriles. It is also used for synthetic API such as anesthetic agents and anti-allergic agents.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.