missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Acetylbenzonitrile, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 167580250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Acetylbenzonitrile | |
98.0 | |
55°C to 58°C | |
98% min. (GC) | |
C9H7NO | |
MFCD00001825 | |
10, 695 | |
NLPHXWGWBKZSJC-UHFFFAOYSA-N | |
4-acetylbenzonitrile | |
74044 | |
98+% |
1443-80-7 | |
100.0 | |
Authentic | |
Glass bottle | |
CH3COC6H4CN | |
25 g | |
4'-cyanoacetophenone, benzonitrile, 4-acetyl, p-cyanoacetophenone, 4-cyanoacetophenone, p-acetylbenzonitrile, 4-acetyl-benzonitrile, benzonitrile, p-acetyl, 4-acetylbenzenecarbonitrile, 4-acetylbenznitrile | |
CC(=O)C1=CC=C(C=C1)C#N | |
145.16 | |
145.16 | |
Crystalline Mass or Crystalline Powder and Chunks |
Chemical Identifiers
1443-80-7 | |
145.16 | |
NLPHXWGWBKZSJC-UHFFFAOYSA-N | |
74044 | |
CC(=O)C1=CC=C(C=C1)C#N |
C9H7NO | |
MFCD00001825 | |
4'-cyanoacetophenone, benzonitrile, 4-acetyl, p-cyanoacetophenone, 4-cyanoacetophenone, p-acetylbenzonitrile, 4-acetyl-benzonitrile, benzonitrile, p-acetyl, 4-acetylbenzenecarbonitrile, 4-acetylbenznitrile | |
4-acetylbenzonitrile |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Obtain special instructions before use.
GHS Signal Word: Warning
EINECSNumber : 215-885-
RUO – Research Use Only