Please login to your online account to display your discounted pricing

4-Acetylbenzenesulfonyl chloride, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  343850050

 View more versions of this product

Catalog No. AC343850050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 218.65
Physical Form Powder and/or Chunks
Quantity 5g
Color White to orange to brown
Melting Point 83° to 87°C
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 1 Danger
Molecular Formula C8H7ClO3S
CAS 1788-10-9
Chemical Name or Material 4-Acetylbenzenesulfonyl chloride
Assay Percent Range 97%
Linear Formula H3CCO(C6H4)SO2Cl
Name Note 98%
MDL Number MFCD00800269
Chloride product may contain varying amounts of inorganic chloride
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Acetylbenzenesulfonyl chloride
Name Note 98%
CAS 1788-10-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H7ClO3S
Linear Formula H3CCO(C6H4)SO2Cl
Formula Weight 218.65
MDL Number MFCD00800269
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents