missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetylbenzenesulfonyl chloride, 97%, ACROS Organics™

$36.28 - $112.96

Chemical Identifiers

CAS 1788-10-9
Molecular Formula C8H7ClO3S
Molecular Weight (g/mol) 218.65
MDL Number MFCD00800269
InChI Key FXVDNCRTKXMSEZ-UHFFFAOYSA-N
Synonym 1-acetyl-4-chlorosulfonyl benzene, 4-acetyl-benzenesulfonyl chloride, 4-acetylbenzene-1-sulfonyl chloride, 4-acetylbenzenesulfonyl chloride, 4-acetylbenzenesulfonylchloride, 4-acetylphenylsulfonyl chloride, acmc-1bs75, benzenesulfonyl chloride, 4-acetyl, benzenesulfonylchloride, 4-acetyl, ksc183k3b
PubChem CID 266418
IUPAC Name 4-acetylbenzenesulfonyl chloride
SMILES CC(=O)C1=CC=C(C=C1)S(=O)(=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC343850010
SDS View Documents
Thermo Fisher Scientific
343850010
1g Glass bottle
Each for $36.28
 
AC343850050
SDS View Documents
Thermo Fisher Scientific
343850050
5g Glass bottle
Each for $112.96
 
AC343850500
SDS View Documents
Thermo Fisher Scientific
AC343850500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1788-10-9
218.65
FXVDNCRTKXMSEZ-UHFFFAOYSA-N
266418
CC(=O)C1=CC=C(C=C1)S(=O)(=O)Cl
C8H7ClO3S
MFCD00800269
1-acetyl-4-chlorosulfonyl benzene, 4-acetyl-benzenesulfonyl chloride, 4-acetylbenzene-1-sulfonyl chloride, 4-acetylbenzenesulfonyl chloride, 4-acetylbenzenesulfonylchloride, 4-acetylphenylsulfonyl chloride, acmc-1bs75, benzenesulfonyl chloride, 4-acetyl, benzenesulfonylchloride, 4-acetyl, ksc183k3b
4-acetylbenzenesulfonyl chloride

Specifications

Authentic
Brown to White
1g
96.0
96% min. (GC)
H3CCO(C6H4)SO2Cl
1-acetyl-4-chlorosulfonyl benzene, 4-acetyl-benzenesulfonyl chloride, 4-acetylbenzene-1-sulfonyl chloride, 4-acetylbenzenesulfonyl chloride, 4-acetylbenzenesulfonylchloride, 4-acetylphenylsulfonyl chloride, acmc-1bs75, benzenesulfonyl chloride, 4-acetyl, benzenesulfonylchloride, 4-acetyl, ksc183k3b
CC(=O)C1=CC=C(C=C1)S(=O)(=O)Cl
218.65
218.65
97%
Glass bottle
83°C to 87°C
1788-10-9
100.0
C8H7ClO3S
MFCD00800269
FXVDNCRTKXMSEZ-UHFFFAOYSA-N
4-acetylbenzenesulfonyl chloride
266418
Powder and/or Chunks
4-Acetylbenzenesulfonyl chloride, 98%
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

SDS
Product Certifications