Please login to your online account to display your discounted pricing

4-Acetylbenzenesulfonyl chloride, 97%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC343850500

 View more versions of this product

Catalog No. AC343850500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder and/or Chunks
Assay ≥96.0%
Formula Weight 218.65
Quantity 50g
Color White to orange to brown
Melting Point 83° to 87°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Acetylbenzenesulfonyl chloride
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 97%
Molecular Formula C8H7ClO3S
Linear Formula H3CCO(C6H4)SO2Cl
MDL Number MFCD00800269
CAS 1788-10-9
Chloride product may contain varying amounts of inorganic chloride
Health Hazard 1 Danger

Specifications

Chemical Name or Material 4-Acetylbenzenesulfonyl chloride
Name Note 98%
CAS 1788-10-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H7ClO3S
Linear Formula H3CCO(C6H4)SO2Cl
Formula Weight 218.65
MDL Number MFCD00800269
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents