Please login to your online account to display your discounted pricing

4-Acetylaminobenzaldehyde thiosemicarbazone, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  336520050

 View more versions of this product

Catalog No. AC336520050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 236.29
Quantity 5g
Merck Index 15, 9442
EINECS Number 203-170-6
RTECS Number AE3850000
CAS Max % 100.0
MDL Number MFCD00022157
CAS 104-06-3
Chemical Name or Material 4-Acetylaminobenzaldehyde thiosemicarbazone
Assay Percent Range 98%
CAS Min % 97.5
Molecular Formula C10H12N4OS
Name Note 98%
Packaging Glass bottle

Specifications

Chemical Name or Material 4-Acetylaminobenzaldehyde thiosemicarbazone
Name Note 98%
CAS 104-06-3
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C10H12N4OS
Formula Weight 236.29
MDL Number MFCD00022157