Please login to your online account to display your discounted pricing

4-Acetylaminobenzaldehyde thiosemicarbazone, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Acetylaminobenzaldehyde thiosemicarbazone
Name Note 98%
CAS 104-06-3
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C10H12N4OS
Formula Weight 236.29
MDL Number MFCD00022157
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC336520050

  SDS
thermo fisher scientific
336520050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.29
Quantity 5g
Merck Index 15, 9442
EINECS Number 203-170-6
RTECS Number AE3850000
CAS Max % 100.0
MDL Number MFCD00022157
CAS 104-06-3
Chemical Name or Material 4-Acetylaminobenzaldehyde thiosemicarbazone
Assay Percent Range 98%
CAS Min % 97.5
Molecular Formula C10H12N4OS
Name Note 98%
Packaging Glass bottle