missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetyl-N-methylaniline, 97%

Manufacturer:  Alfa Aesar™ H5530103

 View more versions of this product

Catalog No. AAH5530103



Specifications

Specifications

4-Acetyl-N-methylaniline
103°C to 107°C
17687-47-7
MFCD09261160
GDBAYICEBDZUNM-UHFFFAOYSA-N
1-[4-(methylamino)phenyl]ethanone
11607936
Solid
Brown to White
1g
C9H11NO
1-4-methylamino phenyl ethan-1-one
CC(=O)C1=CC=C(C=C1)NC
149.193
149.19
97%

Chemical Identifiers

17687-47-7
149.193
GDBAYICEBDZUNM-UHFFFAOYSA-N
11607936
CC(=O)C1=CC=C(C=C1)NC
C9H11NO
MFCD09261160
1-4-methylamino phenyl ethan-1-one
1-[4-(methylamino)phenyl]ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P285-P304+P341-P342+P311-P501c

H334

TSCA : No

Recommended Storage : Ambient temperatures

Documents