missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Acetyl-N-methylaniline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H55301MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Acetyl-N-methylaniline | |
103°C to 107°C | |
C9H11NO | |
250 mg | |
GDBAYICEBDZUNM-UHFFFAOYSA-N | |
1-[4-(methylamino)phenyl]ethanone | |
11607936 | |
97% |
17687-47-7 | |
Brown to White | |
MFCD09261160 | |
4-acetyl-n-methylaniline, 1-4-methylamino phenyl ethanone, 4-acetyl-n-methyl aniline, 1-4-methylamino phenyl ethan-1-one, 1-4-methylamino-phenyl-ethanone, ethanone, 1-4-methylamino phenyl, n-methyl-4-acetylaniline, acmc-1brn7, 4-n-methylaminoacetophenone | |
CNC1=CC=C(C=C1)C(C)=O | |
149.19 | |
149.19 | |
Solid |
Chemical Identifiers
17687-47-7 | |
149.19 | |
GDBAYICEBDZUNM-UHFFFAOYSA-N | |
11607936 | |
CNC1=CC=C(C=C1)C(C)=O |
C9H11NO | |
MFCD09261160 | |
4-acetyl-n-methylaniline, 1-4-methylamino phenyl ethanone, 4-acetyl-n-methyl aniline, 1-4-methylamino phenyl ethan-1-one, 1-4-methylamino-phenyl-ethanone, ethanone, 1-4-methylamino phenyl, n-methyl-4-acetylaniline, acmc-1brn7, 4-n-methylaminoacetophenone | |
1-[4-(methylamino)phenyl]ethanone |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P261-P285-P304+P341-P342+P311-P501c
H334
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only