Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetyl-5-methyl-1-phenyl-1H-pyrazole, 97%

$45.81 - $158.22

Chemical Identifiers

Chemical Name or Material 4-Acetyl-5-methyl-1-phenyl-1H-pyrazole
CAS 6123-63-3
Molecular Formula C12H12N2O
Formula Weight 200.24
MDL Number MFCD00020733
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2080603

  SDS
alfa aesar™
B2080603
1g Each for $45.81

AAB2080606

  SDS
alfa aesar™
B2080606
5g Each for $158.22
Description & Specifications

Specifications

Formula Weight 200.24
Health Hazard 1 Warning
Quantity 1g
Melting Point 103° to 105°C
Assay 97%
MDL Number MFCD00020733
CAS 6123-63-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Acetyl-5-methyl-1-phenyl-1H-pyrazole
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C12H12N2O
Odor Odorless
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.