Please login to your online account to display your discounted pricing

4-Acetyl-4-phenylpiperidine Hydrochloride 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  134580050

 View more versions of this product

Catalog No. AC134580050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 239.74
Assay ≥98.5%
Quantity 5g
Color Slightly yellow to beige
EINECS Number 233-694-0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00039037
Assay Percent Range 99%
CAS Min % 98.5
CAS 10315-03-4
Molecular Formula C13H17NO·ClH
Chemical Name or Material 4-Acetyl-4-phenylpiperidine hydrochloride
Name Note 99%

Specifications

Chemical Name or Material 4-Acetyl-4-phenylpiperidine hydrochloride
Name Note 99%
CAS 10315-03-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13H17NO·ClH
Formula Weight 239.74
MDL Number MFCD00039037