missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetyl-4-phenylpiperidine Hydrochloride 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 134580250

 View more versions of this product

Catalog No. AC134580250


 

Specifications Description & Specifications

Specifications

≥98.5%
239.74
Slightly yellow to beige
99%
4-Acetyl-4-phenylpiperidine hydrochloride
99%
10315-03-4
Glass bottle
Powder
25g
100.0
Authentic
98.5
C13H17NO·ClH
MFCD00039037

Chemical Identifiers

10315-03-4
100.0
C13H17NO·ClH
MFCD00039037
≥98.5%
98.5
99%
239.74
4-Acetyl-4-phenylpiperidine hydrochloride, 99%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 233-694-0