Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetyl-4'-nitrodiphenyl ether, 97%

$71.58 - $284.77

Chemical Identifiers

Chemical Name or Material 4-Acetyl-4'-nitrodiphenyl ether
CAS 75919-92-5
Molecular Formula C14H11NO4
Formula Weight 257.25
MDL Number MFCD00052956
Beilstein 2135124
Synonym 4'-(4-Nitrophenoxy)acetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2059203

  SDS
alfa aesar™
B2059203
1g Each for $71.58

AAB2059206

  SDS
alfa aesar™
B2059206
5g Each for $284.77
Description & Specifications

Specifications

Formula Weight 257.25
Chemical Name or Material 4-Acetyl-4'-nitrodiphenyl ether
Assay 97%
Molecular Formula C14H11NO4
MDL Number MFCD00052956
CAS 75919-92-5
Beilstein 2135124
Synonym 4'-(4-Nitrophenoxy)acetophenone
Melting Point 79° to 81°C
Quantity 1g