Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetyl-4-methylpimelic acid, 97%

$52.19 - $195.49

Chemical Identifiers

Chemical Name or Material 4-Acetyl-4-methylpimelic acid
CAS 19830-09-2
Molecular Formula C10H16O5
Formula Weight 216.24
MDL Number MFCD00052871
Beilstein 1789115
Synonym 4-Acetyl-4-methylheptanedioic acid; 3-Acetyl-3-methylpentane-1,5-dicarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2489106

 
alfa aesar™
B2489106
5g Each for $195.49

AAB2489103

 
alfa aesar™
B2489103
1g Each for $52.19
Description & Specifications

Specifications

Formula Weight 216.24
Chemical Name or Material 4-Acetyl-4-methylpimelic acid
Assay 97%
Beilstein 1789115
Synonym 4-Acetyl-4-methylheptanedioic acid; 3-Acetyl-3-methylpentane-1,5-dicarboxylic acid
Health Hazard 1 Warning
Melting Point 121° to 125°C
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C10H16O5
MDL Number MFCD00052871
CAS 19830-09-2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.