Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetyl-4'-methoxydiphenyl ether, 97%

$113.59 - $418.69

Chemical Identifiers

Chemical Name or Material 4-Acetyl-4'-methoxydiphenyl ether
CAS 54916-28-8
Molecular Formula C15H14O3
Formula Weight 242.28
MDL Number MFCD00052928
Beilstein 2115929
Synonym 4'-(4-Methoxyphenoxy)acetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0982903

 
alfa aesar™
L0982903
1g Each for $113.59

AAL0982906

 
alfa aesar™
L0982906
5g Each for $418.69
Description & Specifications

Specifications

Formula Weight 242.28
Assay 97%
Molecular Formula C15H14O3
MDL Number MFCD00052928
CAS 54916-28-8
Synonym 4'-(4-Methoxyphenoxy)acetophenone
Chemical Name or Material 4-Acetyl-4'-methoxydiphenyl ether
Beilstein 2115929
Melting Point 56° to 58°C
Boiling Point 198° to 201°C (2mmHg)
Quantity 1g