missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetyl-3-fluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

CAS 1351499-39-2
Molecular Formula C14H18BFO3
Molecular Weight (g/mol) 264.103
InChI Key UDNSDVWJDRQRHR-UHFFFAOYSA-N
Synonym 1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone
PubChem CID 66802112
IUPAC Name 1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C(=O)C)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62454MD View Documents Alfa Aesar™
H62454MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6245403 View Documents Alfa Aesar™
H6245403
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1351499-39-2
264.103
1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone
1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
C14H18BFO3
UDNSDVWJDRQRHR-UHFFFAOYSA-N
66802112
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C(=O)C)F

Specifications

250mg
C14H18BFO3
UDNSDVWJDRQRHR-UHFFFAOYSA-N
1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
66802112
Solid
4-Acetyl-3-fluorobenzeneboronic acid pinacol ester
1351499-39-2
1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C(=O)C)F
264.103
264.1
96%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures