missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetyl-3-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6245403

 View more versions of this product

Catalog No. AAH6245403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Acetyl-3-fluorobenzeneboronic acid pinacol ester
1351499-39-2
1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C(=O)C)F
264.103
264.1
96%
1g
C14H18BFO3
UDNSDVWJDRQRHR-UHFFFAOYSA-N
1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
66802112
Solid

Chemical Identifiers

1351499-39-2
264.103
1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone
1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
C14H18BFO3
UDNSDVWJDRQRHR-UHFFFAOYSA-N
66802112
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C(=O)C)F
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents