missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetyl-2-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6225603

 View more versions of this product

Catalog No. AAH6225603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Acetyl-2-fluorobenzeneboronic acid pinacol ester
96%
4-Acetyl-2-fluorophenylboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)C)F
264.103
1g
Solid
1807685-51-3
C14H18BFO3
QRPSPYZFNHVURF-UHFFFAOYSA-N
1-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
75487161
264.1

Chemical Identifiers

1807685-51-3
264.103
4-Acetyl-2-fluorophenylboronic acid pinacol ester
C14H18BFO3
QRPSPYZFNHVURF-UHFFFAOYSA-N
75487161
Documents