Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetoxybiphenyl, 98%

$852.82 - $852.82

Chemical Identifiers

Chemical Name or Material 4-Acetoxybiphenyl
CAS 148-86-7
Molecular Formula C14H12O2
Formula Weight 212.25
MDL Number MFCD00014979
Beilstein 1872019
Synonym 4-Biphenylyl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA121660B

  SDS
alfa aesar™
A121660B
1000g Each for $852.82
Description & Specifications

Specifications

Formula Weight 212.25
Melting Point 85° to 90°C
Boiling Point 196°C
CAS 148-86-7
Synonym 4-Biphenylyl acetate
Chemical Name or Material 4-Acetoxybiphenyl
Assay 98%
Quantity 1000g
Molecular Formula C14H12O2
MDL Number MFCD00014979
Beilstein 1872019