missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetoxybenzonitrile, 97%

Manufacturer:  Alfa Aesar™ H5435803

 View more versions of this product

Catalog No. AAH5435803



Description

Description

Applications
4-Acetoxybenzonitrile is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4-Acetoxybenzonitrile
13031-41-9
MFCD00129158
CJGXWABHYYJNJH-UHFFFAOYSA-N
(4-cyanophenyl) acetate
83062
97%
1g
C9H7NO2
4-acetoxybenzonitrile, 4-acetyloxy benzonitrile, 4-cyanophenyl acetate, benzonitrile, 4-acetyloxy, benzonitrile, p-hydroxy-, acetate ester, eel1pcp8am, p-cyanophenyl acetate, unii-eel1pcp8am
CC(=O)OC1=CC=C(C=C1)C#N
161.16
161.16

Chemical Identifiers

13031-41-9
161.16
CJGXWABHYYJNJH-UHFFFAOYSA-N
83062
CC(=O)OC1=CC=C(C=C1)C#N
C9H7NO2
MFCD00129158
4-acetoxybenzonitrile, 4-acetyloxy benzonitrile, 4-cyanophenyl acetate, benzonitrile, 4-acetyloxy, benzonitrile, p-hydroxy-, acetate ester, eel1pcp8am, p-cyanophenyl acetate, unii-eel1pcp8am
(4-cyanophenyl) acetate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P333+P313-P363-P501c

H302+H312-H317-H319

EINECSNumber : 235-893-8

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents