missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetoxybenzoic acid, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC251451000

 View more versions of this product

Catalog No. AC251451000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Acetoxybenzoic acid
Authentic
Crystalline Powder
99%
CH3COOC6H4COOH
MFCD00002540
189° to 194°C
2345-34-8
Glass bottle
≥98.5%
C9H8O4
180.16
White
100g

Chemical Identifiers

99%
CH3COOC6H4COOH
MFCD00002540
≥98.5%
C9H8O4
180.16
4-Acetoxybenzoic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.