Please login to your online account to display your discounted pricing

4-Acetoxybenzoic acid, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  251450250

 View more versions of this product

Catalog No. AC251450250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Melting Point 189° to 194°C
Quantity 25g
Chemical Name or Material 4-Acetoxybenzoic acid
CAS 2345-34-8
Physical Form White crystalline powder
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C9H8O4
Linear Formula CH3COOC6H4COOH
Formula Weight 180.16g/mol
MDL Number MFCD00002540
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Acetoxybenzoic acid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C9H8O4
Linear Formula CH3COOC6H4COOH
Formula Weight 180.16g/mol
MDL Number MFCD00002540
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

SDS & Safety Documents