Please login to your online account to display your discounted pricing

4-Acetoxybenzoic acid, 99%, ACROS Organics™

$29.51 - $29.51

Chemical Identifiers

Chemical Name or Material 4-Acetoxybenzoic acid
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H8O4
Linear Formula CH3COOC6H4COOH
Formula Weight 180.16
MDL Number MFCD00002540
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC251450250

  SDS
fisher scientific acros
AC251450250
Glass bottle 25g Each for $29.51

AC251451000

  SDS
thermo fisher scientific
AC251451000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC251452500

  SDS
thermo fisher scientific
AC251452500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 180.16
Quantity 25g
Color White
Melting Point 189° to 194°C
Assay Percent Range 99%
MDL Number MFCD00002540
CAS 2345-34-8
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Acetoxybenzoic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C9H8O4
Linear Formula CH3COOC6H4COOH
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.