Learn More
4-Acetoxybenzoic acid, 98+%, Thermo Scientific Chemicals
$57.81 - $308.41
Chemical Identifiers
CAS | 2345-34-8 |
---|---|
Molecular Formula | C9H8O4 |
Molecular Weight (g/mol) | 180.16 |
MDL Number | MFCD00002540 |
InChI Key | GDBUZIKSJGRBJP-UHFFFAOYSA-N |
Synonym | 4-acetoxybenzoic acid, p-acetoxybenzoic acid, 4-acetyloxy benzoic acid, 4-carboxyphenyl acetate, benzoic acid, 4-acetyloxy, p-hydroxybenzoic acid acetate, p-acetyloxybenzoic acid, p-carboxyphenyl acetate, benzoic acid, p-hydroxy-, acetate, 4-acetoxy benzoic acid |
PubChem CID | 16865 |
ChEBI | CHEBI:86560 |
IUPAC Name | 4-acetyloxybenzoic acid |
SMILES | CC(=O)OC1=CC=C(C=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1135014
|
Thermo Scientific Chemicals
A1135014 |
25 g |
Each for $57.81
|
|
|||||
AAA1135018
|
Thermo Scientific Chemicals
A1135018 |
50 g |
Each for $93.51
|
|
|||||
AAA1135030
|
Thermo Scientific Chemicals
A1135030 |
250 g |
Each for $308.41
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2345-34-8 | |
180.16 | |
GDBUZIKSJGRBJP-UHFFFAOYSA-N | |
16865 | |
4-acetyloxybenzoic acid |
C9H8O4 | |
MFCD00002540 | |
4-acetoxybenzoic acid, p-acetoxybenzoic acid, 4-acetyloxy benzoic acid, 4-carboxyphenyl acetate, benzoic acid, 4-acetyloxy, p-hydroxybenzoic acid acetate, p-acetyloxybenzoic acid, p-carboxyphenyl acetate, benzoic acid, p-hydroxy-, acetate, 4-acetoxy benzoic acid | |
CHEBI:86560 | |
CC(=O)OC1=CC=C(C=C1)C(O)=O |
Specifications
2345-34-8 | |
C9H8O4 | |
25 g | |
4-acetoxybenzoic acid, p-acetoxybenzoic acid, 4-acetyloxy benzoic acid, 4-carboxyphenyl acetate, benzoic acid, 4-acetyloxy, p-hydroxybenzoic acid acetate, p-acetyloxybenzoic acid, p-carboxyphenyl acetate, benzoic acid, p-hydroxy-, acetate, 4-acetoxy benzoic acid | |
CC(=O)OC1=CC=C(C=C1)C(O)=O | |
180.16 | |
CHEBI:86560 | |
≥98% |
192°C to 195°C | |
MFCD00002540 | |
1871997 | |
GDBUZIKSJGRBJP-UHFFFAOYSA-N | |
4-acetyloxybenzoic acid | |
16865 | |
180.15 | |
4-Acetoxybenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 219-066-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only