missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetoxybenzoic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1135018

 View more versions of this product

Catalog No. AAA1135018



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Acetoxybenzoic acid
192°C to 195°C
MFCD00002540
1871997
GDBUZIKSJGRBJP-UHFFFAOYSA-N
4-acetyloxybenzoic acid
16865
180.15
2345-34-8
C9H8O4
50 g
4-acetoxybenzoic acid, p-acetoxybenzoic acid, 4-acetyloxy benzoic acid, 4-carboxyphenyl acetate, benzoic acid, 4-acetyloxy, p-hydroxybenzoic acid acetate, p-acetyloxybenzoic acid, p-carboxyphenyl acetate, benzoic acid, p-hydroxy-, acetate, 4-acetoxy benzoic acid
CC(=O)OC1=CC=C(C=C1)C(O)=O
180.16
CHEBI:86560
≥98%

Chemical Identifiers

2345-34-8
180.16
GDBUZIKSJGRBJP-UHFFFAOYSA-N
16865
4-acetyloxybenzoic acid
C9H8O4
MFCD00002540
4-acetoxybenzoic acid, p-acetoxybenzoic acid, 4-acetyloxy benzoic acid, 4-carboxyphenyl acetate, benzoic acid, 4-acetyloxy, p-hydroxybenzoic acid acetate, p-acetyloxybenzoic acid, p-carboxyphenyl acetate, benzoic acid, p-hydroxy-, acetate, 4-acetoxy benzoic acid
CHEBI:86560
CC(=O)OC1=CC=C(C=C1)C(O)=O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 219-066-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only