missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetoxybenzoic acid, 98+%

Manufacturer:  Alfa Aesar™ A1135030

 View more versions of this product

Catalog No. AAA1135030


 

Specifications

Specifications

4-Acetoxybenzoic acid
≥98%
MFCD00002540
GDBUZIKSJGRBJP-UHFFFAOYSA-N
4-acetyloxybenzoic acid
16865
180.15
250g
2345-34-8
C9H8O4
1871997
CC(=O)OC1=CC=C(C=C1)C(=O)O
180.159
CHEBI:86560
192°C to 195°C

Chemical Identifiers

2345-34-8
180.159
GDBUZIKSJGRBJP-UHFFFAOYSA-N
CHEBI:86560
C9H8O4
MFCD00002540
16865
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents