Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetoxybenzaldehyde, 98%

$1,069.80 - $1,069.80

Chemical Identifiers

Chemical Name or Material 4-Acetoxybenzaldehyde
CAS 878-00-2
Molecular Formula C9H8O3
Formula Weight 164.16
MDL Number MFCD00003384
Beilstein 1101359
Synonym 4-Formylphenyl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2490722

 
alfa aesar™
B2490722
100g Each for $1,069.80
Description & Specifications

Specifications

Formula Weight 164.16
Chemical Name or Material 4-Acetoxybenzaldehyde
TSCA TSCA
Assay 98%
Molecular Formula C9H8O3
MDL Number MFCD00003384
CAS 878-00-2
Beilstein 1101359
Synonym 4-Formylphenyl acetate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 170° to 172°C (35mmHg)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Quantity 100g

Applications
4-Acetoxybenzaldehyde is used as pharmaceutical intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.