missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4'-Acetoxyacetophenone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2228414
Description
4'-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.
Solubility
Slightly soluble in water(4450 mg/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
4'-Acetoxyacetophenone | |
51°C to 54°C | |
C10H10O3 | |
25 g | |
4'-acetoxyacetophenone, p-acetoxyacetophenone, 4-acetoxyacetophenone, p-acetylphenyl acetate, 4-acetylphenyl acetate, unii-eo1xk390jf, ethanone, 1-4-acetyloxy phenyl, 4-acetoxy acetophenone, 4-acetoxy-acetophenone, eo1xk390jf | |
SMIOEQSLJNNKQF-UHFFFAOYSA-N | |
(4-acetylphenyl) acetate | |
83063 | |
178.19 |
13031-43-1 | |
165°C to 170°C (14 mmHg) | |
MFCD00017229 | |
2093080 | |
Slightly soluble in water(4450mg/L). | |
CC(=O)OC1=CC=C(C=C1)C(C)=O | |
178.19 | |
CHEBI:86558 | |
99% |
Chemical Identifiers
13031-43-1 | |
178.19 | |
SMIOEQSLJNNKQF-UHFFFAOYSA-N | |
83063 | |
(4-acetylphenyl) acetate |
C10H10O3 | |
MFCD00017229 | |
4'-acetoxyacetophenone, p-acetoxyacetophenone, 4-acetoxyacetophenone, p-acetylphenyl acetate, 4-acetylphenyl acetate, unii-eo1xk390jf, ethanone, 1-4-acetyloxy phenyl, 4-acetoxy acetophenone, 4-acetoxy-acetophenone, eo1xk390jf | |
CHEBI:86558 | |
CC(=O)OC1=CC=C(C=C1)C(C)=O |
Safety and Handling
EINECSNumber : 235-894-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only