missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetoxy-3-methoxybenzaldehyde, 98%

Manufacturer:  Alfa Aesar™ A1210330

 View more versions of this product

Catalog No. AAA1210330



Specifications

Specifications

4-Acetoxy-3-methoxybenzaldehyde
250g
C10H10O4
1963795
PZSJOBKRSVRODF-UHFFFAOYSA-N
(4-formyl-2-methoxyphenyl) acetate
61229
194.18
74°C to 78°C
881-68-5
MFCD00003362
vanillin acetate, 4-acetoxy-3-methoxybenzaldehyde, acetovanillin, acetylvanillin, vanillin, acetate, 4-o-acetylvanillin, o-acetylvanillin, benzaldehyde, 4-acetyloxy-3-methoxy, 3-methoxy-4-acetoxybenzaldehyde, acetyl vanillin
CC(=O)OC1=C(C=C(C=C1)C=O)OC
194.186
CHEBI:86956
98%

Chemical Identifiers

881-68-5
194.186
PZSJOBKRSVRODF-UHFFFAOYSA-N
61229
(4-formyl-2-methoxyphenyl) acetate
C10H10O4
MFCD00003362
vanillin acetate, 4-acetoxy-3-methoxybenzaldehyde, acetovanillin, acetylvanillin, vanillin, acetate, 4-o-acetylvanillin, o-acetylvanillin, benzaldehyde, 4-acetyloxy-3-methoxy, 3-methoxy-4-acetoxybenzaldehyde, acetyl vanillin
CHEBI:86956
CC(=O)OC1=C(C=C(C=C1)C=O)OC
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents