Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetoxy-3-ethoxybenzaldehyde, 99%

$94.97 - $341.72

Chemical Identifiers

Chemical Name or Material 4-Acetoxy-3-ethoxybenzaldehyde
CAS 72207-94-4
Molecular Formula C11H12O4
Formula Weight 208.22
MDL Number MFCD01666413
Synonym 2-Ethoxy-4-formylphenyl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5048803

 
alfa aesar™
H5048803
1g Each for $341.72

AAH50488MD

 
alfa aesar™
H50488MD
250mg Each for $94.97
Description & Specifications

Specifications

Formula Weight 208.22
Chemical Name or Material 4-Acetoxy-3-ethoxybenzaldehyde
TSCA TSCA
Molecular Formula C11H12O4
MDL Number MFCD01666413
CAS 72207-94-4
Synonym 2-Ethoxy-4-formylphenyl acetate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number CU4381000
Assay 99%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
4-Acetoxy-3-ethoxybenzaldehyde is used in organic synthesis. It is used as catalytic agent; petrochemical additive.

Solubility
Soluble in water (986.4 mg / L) at 25°C.

Notes
Air Sensitive. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.