Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetoxy-2,5-dimethyl-3(2H)-furanone, 98%

$101.66 - $101.66

Chemical Identifiers

Chemical Name or Material 4-Acetoxy-2,5-dimethyl-3(2H)-furanone
CAS 4166-20-5
Molecular Formula C8H10O4
Formula Weight 170.17g/mol
MDL Number MFCD00799462
Synonym Furaneol acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2207909

 
alfa aesar™
B22079-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2207918

 
alfa aesar™
B22079-18
50g Each for $101.66
Description & Specifications

Specifications

Boiling Point 96° to 98°C (1mmHg)
Quantity 10g
Chemical Name or Material 4-Acetoxy-2,5-dimethyl-3(2H)-furanone
CAS 4166-20-5
Assay 98%
Molecular Formula C8H10O4
Formula Weight 170.17g/mol
MDL Number MFCD00799462
Synonym Furaneol acetate