missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetamidothiophenol 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 102340250

 View more versions of this product

Catalog No. AC102340250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥94%
Powder
Yellow
149° to 154°C
94.0
p-Mercaptoacetanilide
Glass bottle
95%
C8H9NOS
4-Acetamidothiophenol
167.23
25g
13, 541
100.0
Authentic
CH3CONHC6H4SH
1126-81-4
95%
MFCD00004848

Chemical Identifiers

1126-81-4
100.0
C8H9NOS
167.23
p-Mercaptoacetanilide
≥94%
94.0
95%
CH3CONHC6H4SH
MFCD00004848
4-Acetamidothiophenol, 95%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 214-427-7