Please login to your online account to display your discounted pricing

4-Acetamidothiophenol 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  102340050

 View more versions of this product

Catalog No. AC102340050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 167.23
Physical Form Powder
Quantity 5g
Color Yellow
EINECS Number 214-427-7
Melting Point 149° to 154°C
Beilstein 13, 541
CAS Max % 100.0
CAS Min % 94.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Linear Formula CH3CONHC6H4SH
CAS 1126-81-4
Name Note 95%
MDL Number MFCD00004848
Assay Percent Range 95%
Chemical Name or Material 4-Acetamidothiophenol
Molecular Formula C8H9NOS
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym p-Mercaptoacetanilide

Specifications

Chemical Name or Material 4-Acetamidothiophenol
Name Note 95%
CAS 1126-81-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H9NOS
Linear Formula CH3CONHC6H4SH
Formula Weight 167.23
MDL Number MFCD00004848
Synonym p-Mercaptoacetanilide
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents