Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetamidopiperidine, 97%

$61.26 - $61.26

Chemical Identifiers

Chemical Name or Material 4-Acetamidopiperidine
CAS 5810-56-0
Molecular Formula C7H14N2O
Formula Weight 142.2
MDL Number MFCD05254805
Synonym N-(4-Piperidinyl)acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50981MD

 
alfa aesar™
H50981MD
250mg Each for $61.26
Description & Specifications

Specifications

Formula Weight 142.2
MDL Number MFCD05254805
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Acetamidopiperidine
Assay 97%
Molecular Formula C7H14N2O
CAS 5810-56-0
Synonym N-(4-Piperidinyl)acetamide
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 141°C
Quantity 250mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.