Learn More
4-Acetamidophenylboronic acid, 97+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 367610050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Acetamidophenylboronic acid | |
97.0 | |
Authentic | |
Glass bottle | |
MFCD02179451 | |
4-acetylaminophenylboronic acid, 4-acetamidophenyl boronic acid, p-acetamidophenylboronic acid, 4-acetamidobenzeneboronic acid, 4-acetamidophenyl-boronic acid, 4-acetaminophenyl boronic acid, 4-acetylaminophenyl boronic acid, 4-acetylamino phenylboronic acid, boronic acid, 4-acetylamino phenyl | |
VYEWTHXZHHATTA-UHFFFAOYSA-N | |
(4-acetamidophenyl)boronic acid | |
2734657 | |
97+% |
101251-09-6 | |
100.0 | |
97% min. (GC) | |
C8H10BNO3 | |
5g | |
Solubility in water: 85g/L (70°C). Other solubilities: slightly soluble in ethylacetate,soluble in meoh,etoh,dmso | |
B(C1=CC=C(C=C1)NC(=O)C)(O)O | |
178.98 | |
178.98 |
Chemical Identifiers
101251-09-6 | |
178.98 | |
VYEWTHXZHHATTA-UHFFFAOYSA-N | |
2734657 | |
B(C1=CC=C(C=C1)NC(=O)C)(O)O |
C8H10BNO3 | |
MFCD02179451 | |
4-acetylaminophenylboronic acid, 4-acetamidophenyl boronic acid, p-acetamidophenylboronic acid, 4-acetamidobenzeneboronic acid, 4-acetamidophenyl-boronic acid, 4-acetaminophenyl boronic acid, 4-acetylaminophenyl boronic acid, 4-acetylamino phenylboronic acid, boronic acid, 4-acetylamino phenyl | |
(4-acetamidophenyl)boronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
Recommended Storage : Refrigerator +4°C
RUO â Research Use Only