missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetamidophenol, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 102332500

 View more versions of this product

Catalog No. AC102332500


 

Specifications Description & Specifications

Specifications

≥97.5% (HPLC)
151.16
250g
1.2930g/mL
Authentic
CH3CONHC6H4OH
98%
97.5
Plastic bottle
168° to 172°C
Crystals or Crystalline Powder
White
100.0
APAP
4-Acetamidophenol
C8H9NO2
103-90-2

Chemical Identifiers

103-90-2
100.0
C8H9NO2
151.16
4-Acetamidophenol
97.5
98%
CH3CONHC6H4OH
APAP
≥97.5% (HPLC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning