missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetamidophenol, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC102330051

 View more versions of this product

Catalog No. AC102330051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Acetamidophenol
Authentic
1.293
168.0°C to 172.0°C
103-90-2
100.0
C8H9NO2
13,46
4-acetamidophenol
RZVAJINKPMORJF-UHFFFAOYSA-N
N-(4-hydroxyphenyl)acetamide
1983
151.16
98%
1.2930g/mL
Plastic bottle
White
5kg
97.5
98%
CH3CONHC6H4OH
15,47
Solubility in water: soluble 14g/L in water (20°C). Other solubilities: soluble in ethanol,acetone,alcohol,methanol,,dmf,ethylene dichloride,ethyl acetate,practically insoluble in petr.ether,pentane and,benzene
CC(=O)NC1=CC=C(C=C1)O
151.16
CHEBI:46195
Crystals or Crystalline Powder

Chemical Identifiers

103-90-2
151.16
4-acetamidophenol
CHEBI:46195
CC(=O)NC1=CC=C(C=C1)O
C8H9NO2
RZVAJINKPMORJF-UHFFFAOYSA-N
1983
N-(4-hydroxyphenyl)acetamide
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents