Learn More
4-Acetamidophenol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1124036
Description
Cyclooxygenase inhibitor and widely used analgesic. 4-Acetamidophenol formulated pharmaceutical products are used as antiinfectant, analgesic, anti rheumatic and antipyretic. It is used as an intermediate in organic synthesis, hydrogen peroxide stabilizer and photographic chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclooxygenase inhibitor and widely used analgesic. 4-Acetamidophenol formulated pharmaceutical products are used as antiinfectant, analgesic, anti rheumatic and antipyretic. It is used as an intermediate in organic synthesis, hydrogen peroxide stabilizer and photographic chemicals.
Solubility
Soluble in alcohol, dimethylformamide, ethylene dichloride, acetone and ethyl acetate. Slightly soluble in water and ether.
Notes
Incompatible with strong oxidizing agents, acids and alkali metals.
Specifications
4-Acetamidophenol | |
168°C to 172°C | |
C8H9NO2 | |
500 g | |
14,47 | |
Soluble in alcohol,dimethylformamide,ethylene dichloride,acetone and ethyl acetate. Slightly soluble in water and ether. | |
CC(=O)NC1=CC=C(O)C=C1 | |
151.17 | |
CHEBI:46195 | |
98% |
103-90-2 | |
1.293 | |
MFCD00002328 | |
2208089 | |
acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol | |
RZVAJINKPMORJF-UHFFFAOYSA-N | |
N-(4-hydroxyphenyl)acetamide | |
1983 | |
151.17 |
Chemical Identifiers
103-90-2 | |
151.17 | |
RZVAJINKPMORJF-UHFFFAOYSA-N | |
1983 | |
N-(4-hydroxyphenyl)acetamide |
C8H9NO2 | |
MFCD00002328 | |
acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol | |
CHEBI:46195 | |
CC(=O)NC1=CC=C(O)C=C1 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 203-157-5
RTECSNumber : AE4200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only