Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetamidobutyric acid, 99%

$626.49 - $2,850.74

Chemical Identifiers

Chemical Name or Material 4-Acetamidobutyric acid
CAS 3025-96-5
Molecular Formula CH3CONH(CH2)3CO2H
Formula Weight 145.16
MDL Number MFCD00020552
Beilstein 1762288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1103918

  SDS
alfa aesar™
A1103918
50g Each for $626.49

AAA1103930

  SDS
alfa aesar™
A1103930
250g Each for $2,850.74
Description & Specifications

Specifications

Formula Weight 145.16
Chemical Name or Material 4-Acetamidobutyric acid
Assay 99%
Molecular Formula CH3CONH(CH2)3CO2H
MDL Number MFCD00020552
CAS 3025-96-5
Beilstein 1762288
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 129° to 131°C
Quantity 50g
RTECS Number ES5610000

Applications
4-Acetamidobutyric acid is used as pharmaceutical intermediate.

Solubility
Soluble in water (19.9 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.