missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetamidobenzenethiol 90.0+%, TCI America™

$106.05 - $287.22

Chemical Identifiers

CAS 1126-81-4
Molecular Formula C8H9NOS
Molecular Weight (g/mol) 167.23
MDL Number MFCD00004848
InChI Key AYEQJLOHMLYKAV-UHFFFAOYSA-N
Synonym 4'-mercaptoacetanilide, 4-acetamido thiophenol, 4-acetamidobenzenethiol, 4-acetamidothiophenol, 4-acetaminothiophenol, 4-acetylaminothiophenol, 4-mercaptoacetanilide, acetanilide, 4'-mercapto, n-4-mercaptophenyl acetamide, n-4-sulfanylphenyl acetamide
PubChem CID 70777
IUPAC Name N-(4-sulfanylphenyl)acetamide
SMILES CC(=O)NC1=CC=C(S)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A19095G
View Documents
Tci America
A19095G
5g
Each for $106.05
 
A190925G
View Documents
Tci America
A190925G
25g
Each for $287.22
 
Description

Description

Specifications

Chemical Identifiers

1126-81-4
167.23
AYEQJLOHMLYKAV-UHFFFAOYSA-N
70777
CC(=O)NC1=CC=C(S)C=C1
C8H9NOS
MFCD00004848
4'-mercaptoacetanilide, 4-acetamido thiophenol, 4-acetamidobenzenethiol, 4-acetamidothiophenol, 4-acetaminothiophenol, 4-acetylaminothiophenol, 4-mercaptoacetanilide, acetanilide, 4'-mercapto, n-4-mercaptophenyl acetamide, n-4-sulfanylphenyl acetamide
N-(4-sulfanylphenyl)acetamide

Specifications

Yellow
5g
C8H9NOS
4'-mercaptoacetanilide, 4-acetamido thiophenol, 4-acetamidobenzenethiol, 4-acetamidothiophenol, 4-acetaminothiophenol, 4-acetylaminothiophenol, 4-mercaptoacetanilide, acetanilide, 4'-mercapto, n-4-mercaptophenyl acetamide, n-4-sulfanylphenyl acetamide
CC(=O)NC1=CC=C(S)C=C1
167.23
167.23
≥90.0% (T)
153°C
1126-81-4
MFCD00004848
AYEQJLOHMLYKAV-UHFFFAOYSA-N
N-(4-sulfanylphenyl)acetamide
70777
Crystal-Powder at 20°C
4-Acetamidobenzenethiol
Safety and Handling

Safety and Handling

TSCA : No

SDS
Product Certifications