missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetamidobenzenesulfonyl Chloride 98.0+%, TCI America™

$116.11 - $336.13

Chemical Identifiers

CAS 121-60-8
Molecular Formula C8H8ClNO3S
Molecular Weight (g/mol) 233.666
MDL Number MFCD00007442
InChI Key GRDXCFKBQWDAJH-UHFFFAOYSA-N
Synonym n-acetylsulfanilyl chloride, 4-acetamidobenzene-1-sulfonyl chloride, dagenan chloride, 4-acetylamino benzenesulfonyl chloride, 4-acetamidophenylsulfonyl chloride, benzenesulfonyl chloride, 4-acetylamino, acetylsulfanilyl chloride, p-acetamidobenzenesulfonyl chloride, n-acetylsulphanilyl chloride, p-acetaminobenzenesulfonyl chloride
PubChem CID 8481
IUPAC Name 4-acetamidobenzenesulfonyl chloride
SMILES CC(=O)NC1=CC=C(C=C1)S(=O)(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A007425G View Documents TCI America
A007425G
25g Each for $116.11
 
A0074100G View Documents TCI America
A0074100G
100g Each for $336.13
 
Specifications

Chemical Identifiers

121-60-8
233.666
GRDXCFKBQWDAJH-UHFFFAOYSA-N
8481
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)Cl
C8H8ClNO3S
MFCD00007442
n-acetylsulfanilyl chloride, 4-acetamidobenzene-1-sulfonyl chloride, dagenan chloride, 4-acetylamino benzenesulfonyl chloride, 4-acetamidophenylsulfonyl chloride, benzenesulfonyl chloride, 4-acetylamino, acetylsulfanilyl chloride, p-acetamidobenzenesulfonyl chloride, n-acetylsulphanilyl chloride, p-acetaminobenzenesulfonyl chloride
4-acetamidobenzenesulfonyl chloride

Specifications

White-Yellow
25g
C8H8ClNO3S
3261
GRDXCFKBQWDAJH-UHFFFAOYSA-N
4-acetamidobenzenesulfonyl chloride
8481
Crystal-Powder at 20°C
4-Acetamidobenzenesulfonyl Chloride
146°C
121-60-8
MFCD00007442
n-acetylsulfanilyl chloride, 4-acetamidobenzene-1-sulfonyl chloride, dagenan chloride, 4-acetylamino benzenesulfonyl chloride, 4-acetamidophenylsulfonyl chloride, benzenesulfonyl chloride, 4-acetylamino, acetylsulfanilyl chloride, p-acetamidobenzenesulfonyl chloride, n-acetylsulphanilyl chloride, p-acetaminobenzenesulfonyl chloride
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)Cl
233.666
233.67
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1988

RTECSNumber : DB8837500

TSCA : Yes