Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetamidobenzenesulfonyl chloride, 98+%

Chemical Identifiers

Chemical Name or Material 4-Acetamidobenzenesulfonyl chloride
CAS 121-60-8
Molecular Formula C8H8ClNO3S
Formula Weight 233.67
MDL Number MFCD00007442
UN Number UN3261
Beilstein 746676
Merck Index 14,103
Synonym N-Acetylsulfanilyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA115110B

  SDS
alfa aesar™
A115110B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 233.67
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Quantity 1000g
TSCA TSCA
Assay ≥98%
MDL Number MFCD00007442
CAS 121-60-8
Molecular Formula C8H8ClNO3S
RTECS Number DB8837500
Chemical Name or Material 4-Acetamidobenzenesulfonyl chloride
UN Number UN3261
Beilstein 746676
Melting Point ≈146°C (dec.)
Synonym N-Acetylsulfanilyl chloride
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Merck Index 14,103
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
N-Acetylsulfanilyl chloride is used in the preparation of sulfanilamide and its derivatives which are intermediates to produce sulfa drugs. They are used in the prevention and treatment of bacterial infections, diabetes mellitus, edema, hypertension, and gout. It is also used as a Pharma raw material. N-Acetylsulfanilyl chloride widely used in the fields of dye, medicine, mainly used for preparation of Sulfanilamide, Sulfanilylureal, Sulfatolamide, Sulphathiourea , Sulfaguanidine and Sulfacetamide and etc.

Solubility
It is slightly in water. But soluble in benzene, chloroform and ether.

Notes
Stable under ordinary conditions. Moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents, Strong bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.