missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetamidoantipyrine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1500614

 View more versions of this product

Catalog No. AAA1500614



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Acetamidoantipyrine
200°C to 203°C
MFCD00003141
Light sensitive
OIAGWXKSCXPNNZ-UHFFFAOYSA-N
N-(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)acetamide
65743
245.28
83-15-8
C13H15N3O2
25 g
4-acetamidoantipyrine, 4-acetaminoantipyrine, acetamidoantipyrine, acetylaminoantipyrine, 4-acetoaminoantipyrine, 4-acetylaminophenazone, 4-acetylaminoantipyrine, antipyrine, 4-acetamido, n-acetyl-4-aminoantipyrine, acetylated 4-aminoantipyrine
CC1=C(C(=O)N(N1C)C2=CC=CC=C2)NC(=O)C
245.282
CHEBI:83513
97%

Chemical Identifiers

83-15-8
245.282
OIAGWXKSCXPNNZ-UHFFFAOYSA-N
65743
N-(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)acetamide
C13H15N3O2
MFCD00003141
4-acetamidoantipyrine, 4-acetaminoantipyrine, acetamidoantipyrine, acetylaminoantipyrine, 4-acetoaminoantipyrine, 4-acetylaminophenazone, 4-acetylaminoantipyrine, antipyrine, 4-acetamido, n-acetyl-4-aminoantipyrine, acetylated 4-aminoantipyrine
CHEBI:83513
CC1=C(C(=O)N(N1C)C2=CC=CC=C2)NC(=O)C
Safety and Handling

Safety and Handling

EINECSNumber : 201-457-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only