Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetamido-9-fluorenone, 99%

$66.15 - $66.15

Chemical Identifiers

Chemical Name or Material 4-Acetamido-9-fluorenone
CAS 42135-35-3
Molecular Formula C15H11NO2
Formula Weight 237.26
MDL Number MFCD00010782
Synonym 4-Acetylamino-9-fluorenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2241403

  SDS
alfa aesar™
B2241403
1g Each for $66.15
Description & Specifications

Specifications

Formula Weight 237.26
Chemical Name or Material 4-Acetamido-9-fluorenone
CAS 42135-35-3
Melting Point 262° to 264°C
Synonym 4-Acetylamino-9-fluorenone
Molecular Formula C15H11NO2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
MDL Number MFCD00010782
Assay 99%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.